Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Cynomolgus CD24, Fc Tag, primary amine labeling CD4-C8254 is expressed from human 293 cells HEK293. It contains AA Thr 27 - Lys 54 Accession I7GKK1-1. 1 * 25 µG
Catalog # ACRBCD4-C8254-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD24, Fc Tag, primary amine labeling CD4-C8254 is expressed from human 293 cells HEK293. It contains AA Thr 27 - Lys 54 Accession I7GKK1-1. 1 * 25 µG
Catalog # ACRBCD4-C8254-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together