ScienceLine plus pH combination electrode Micro electrode with plug head, glass shaft, Pt junction, electrolyte KCl 3 mol/l, cylindrical membrane, A glass, length 70/130 mm, 12/5 mm Ø, -5...+100 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285101330
Dodávateľ: SI ANALYTICS
Frequently Bought Together