Services for equipment
Preventive maintenance, operational testing and calibration of vacuum oven at customer site. 1× Temperature and Vacuum Calibration (1 probe). Calibration Certificate included. Spare parts and travel fees not included
Catalog # SERW2220.1131.01
Supplier: AVANTOR SERVICES
Jump to:
Product Details & Documents
Specifications
- Description:Preventive maintenance, operational testing and calibration of vacuum oven at customer site. 1× Temperature and Vacuum Calibration (1 probe). Calibration Certificate included. Spare parts and travel fees not included
- Seq:46