Order Entry
Slovakia
ContactUsLinkComponent
Cover glasses No. 1.5, 16 mm dia., 100 pcs./two-part-box 1 * 1.000 items
Catalog # MARI0112560
Supplier:  MARIENFELD
Cover glasses No. 1.5, 16 mm dia., 100 pcs./two-part-box 1 * 1.000 items
Catalog # MARI0112560
Supplier:  MARIENFELD
Frequently Bought Together