Reagecon Standard Colour Solution OY (Orange/Yellow) according to Chinese Pharmacopoeia (ChP) 1 * 100 mL
Catalog # REGECPSRSOY01
Supplier: REAGECON
Reagecon Standard Colour Solution OY (Orange/Yellow) according to Chinese Pharmacopoeia (ChP) 1 * 100 mL
Catalog # REGECPSRSOY01
Supplier: REAGECON
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dextrose, Reference Standard
Supplier: USP
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: METTLER TOLEDO
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Titration beakers
Supplier: METTLER TOLEDO
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals