Order Entry
Slovakia
ContactUsLinkComponent
SARS S1 protein, His Tag SARS S1 protein, His Tag (S1N-S52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 14 - Ar g 667 (Accession AAP13567.1). This protein carries a polyhistidine tag at the C-terminus. The protein has a calcula k 1 * 100 µG
Catalog # ACRBS1NS52H5100UG
Supplier:  ACRO Biosystems
SARS S1 protein, His Tag SARS S1 protein, His Tag (S1N-S52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 14 - Ar g 667 (Accession AAP13567.1). This protein carries a polyhistidine tag at the C-terminus. The protein has a calcula k 1 * 100 µG
Catalog # ACRBS1NS52H5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together