Order Entry
Slovakia
ContactUsLinkComponent
2019-nCoV (COVID-19) Envelope protein, His Tag 2019-nCoV (COVID-19) Envelope protein, His Tag (ENN-C5128) is expressed from E.coli c ells. It contains AA Met 1 -Val 75 (Accession QHU79206.1). This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBENNC5128100UG
Supplier:  ACRO Biosystems
2019-nCoV (COVID-19) Envelope protein, His Tag 2019-nCoV (COVID-19) Envelope protein, His Tag (ENN-C5128) is expressed from E.coli c ells. It contains AA Met 1 -Val 75 (Accession QHU79206.1). This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBENNC5128100UG
Supplier:  ACRO Biosystems
Frequently Bought Together