Order Entry
Slovakia
ContactUsLinkComponent
2019-nCoV (COVID-19) S2 protein, His Tag 2019-nCoV (COVID-19) S2 protein, His Tag (S2N-C52H5) is expressed from human 293 cells (HEK 293). It contains AA Ser 686 - Pro1213 (Accession QHD43416.1). This protein carries a polyhistidine tag at the C-te t 1 * 1 mg
Catalog # ACRBS2NC52H51MG
Supplier:  ACRO Biosystems
2019-nCoV (COVID-19) S2 protein, His Tag 2019-nCoV (COVID-19) S2 protein, His Tag (S2N-C52H5) is expressed from human 293 cells (HEK 293). It contains AA Ser 686 - Pro1213 (Accession QHD43416.1). This protein carries a polyhistidine tag at the C-te t 1 * 1 mg
Catalog # ACRBS2NC52H51MG
Supplier:  ACRO Biosystems
Frequently Bought Together