Recombinat Protein,Human CD99 MIC2 Protein, Fc Tag,Human CD99, Fc Tag (CD9-H5257) is expressed from human 293 cells (HEK293). It con tains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 100 µG
Catalog # ACRBCD9-H5257100UG
Supplier: ACRO Biosystems
Frequently Bought Together