Order Entry
Slovakia
ContactUsLinkComponent
ActiveMax® Human Oncostatin M, Tag Free (OSM-H5213) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Arg 252 (Acc ession NP065391.1). 1 * 50 µG
Catalog # ACRBOSM-H5213-50UG
Supplier:  ACRO Biosystems
ActiveMax® Human Oncostatin M, Tag Free (OSM-H5213) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Arg 252 (Acc ession NP065391.1). 1 * 50 µG
Catalog # ACRBOSM-H5213-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together