Order Entry
Slovakia
ContactUsLinkComponent
ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 136 (Accession NP001165097)
  ACRBVE0-H5212-20UG
ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - A rg 136 (Accession NP001165097)
  ACRBVE0-H5212-20UG
Frequently Bought Together