Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
Catalog # ACRBFA2-H5149-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human FABP2 I-FABP Protein, His Tag,Human FABP2, His Tag (FA2-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 132 (Accession NP000125).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
Catalog # ACRBFA2-H5149-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together