Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human IL-17 RA, His,Avitag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Acc ession Q96F46-1). 1 * 25 µG
  ACRBILR-H82E5-25UG
Biotinylated Human IL-17 RA, His,Avitag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Acc ession Q96F46-1). 1 * 25 µG
  ACRBILR-H82E5-25UG
Frequently Bought Together