Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human M-CSF R, Avitag,His Tag (CSR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 ( Accession P07333-1). 1 * 200 µG
Catalog # ACRBCSR-H82E0200UG
Supplier:  ACRO Biosystems
Biotinylated Human M-CSF R, Avitag,His Tag (CSR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 ( Accession P07333-1). 1 * 200 µG
Catalog # ACRBCSR-H82E0200UG
Supplier:  ACRO Biosystems
Frequently Bought Together