Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accessio n Q95LI5). 1 * 1 mg
Catalog # ACRBCDE-C5254-1MG
Supplier:  ACRO Biosystems
Cynomolgus CD3 epsilon, Fc,His Tag (CDE-C5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accessio n Q95LI5). 1 * 1 mg
Catalog # ACRBCDE-C5254-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together