Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (A ccession ALV66454.1). 1 * 100 µG
Catalog # ACRBLA1-C52A3100UG
Supplier:  ACRO Biosystems
Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (A ccession ALV66454.1). 1 * 100 µG
Catalog # ACRBLA1-C52A3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together