Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse BCMA, Fc,Avitag (BCA-M82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession O88472-1). 1 * 25 µG
Catalog # ACRBBCA-M82F0-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse BCMA, Fc,Avitag (BCA-M82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession O88472-1). 1 * 25 µG
Catalog # ACRBBCA-M82F0-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together