Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus LILRB4, His Tag (CDK-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP0152 97198.1). 1 * 100 µG
Katalógové číslo ACRBCDK-C5227100UG
Supplier:  ACRO Biosystems
Cynomolgus LILRB4, His Tag (CDK-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession XP0152 97198.1). 1 * 100 µG
Katalógové číslo ACRBCDK-C5227100UG
Supplier:  ACRO Biosystems
Frequently Bought Together