Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acce ssion P35916-1). 1 * 200 µG
Catalog # ACRBFL4-H82E1200UG
Supplier:  ACRO Biosystems
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Acce ssion P35916-1). 1 * 200 µG
Catalog # ACRBFL4-H82E1200UG
Supplier:  ACRO Biosystems
Frequently Bought Together