Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (FC6-C82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 1 7 - Gln 208 (Accession Q8SPW2-1). 1 * 200 µG
Catalog # ACRBFC6-C82E0200UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD16, His,Avitag (BLI verified) (FC6-C82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 1 7 - Gln 208 (Accession Q8SPW2-1). 1 * 200 µG
Catalog # ACRBFC6-C82E0200UG
Supplier:  ACRO Biosystems
Frequently Bought Together