Recombinat Protein,Human Carbonic Anhydrase II CA2 Protein, His Tag,Human CA2, His Tag (CA2-H5228) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Lys 260 (Accession NP000058.1).This protein carries a polyhistidine tag at the C-te 1 * 1 mg
Catalog # ACRBCA2-H5228-1MG
Supplier: ACRO Biosystems
Frequently Bought Together