Biotinylated Human CD64, His,Avitag (SPR BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 µG
Catalog # ACRBFCA-H82E8-25UG
Supplier: ACRO Biosystems
Biotinylated Human CD64, His,Avitag (SPR BLI verified) (FCA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gln 1 6 - Pro 288 (Accession P12314-1). 1 * 25 µG
Catalog # ACRBFCA-H82E8-25UG
Supplier: ACRO Biosystems
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Reference electrode filling solutions
Supplier: METTLER TOLEDO
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Defined foetal bovine serum, US origin, HyClone™
Supplier: HyClone products (Cytiva)
Human recombinant Fc gamma RIIIB / CD16b (NA1) (from HEK293)
Supplier: ACRO Biosystems
Storage vials, depyrogenated glass
Supplier: EP SCIENTIFIC