Order Entry
Slovakia
ContactUsLinkComponent
Human TIGIT, Mouse IgG2a Fc Tag, low endotoxin (TIT-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 1 41 (Accession Q495A1-1). 1 * 100 µG
Catalog # ACRBTIT-H5253100UG
Supplier:  ACRO Biosystems
Human TIGIT, Mouse IgG2a Fc Tag, low endotoxin (TIT-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 1 41 (Accession Q495A1-1). 1 * 100 µG
Catalog # ACRBTIT-H5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together