Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBPTD-H52H9-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBPTD-H52H9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together