Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human B7-H7, Fc,Avitag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accessi on Q9UM44-1). 1 * 25 µG
Catalog # ACRBB77-H82F5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H7, Fc,Avitag (B77-H82F5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accessi on Q9UM44-1). 1 * 25 µG
Catalog # ACRBB77-H82F5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together