Order Entry
Slovakia
ContactUsLinkComponent
Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 100 µG
Catalog # ACRBTN1-H5251100UG
Supplier:  ACRO Biosystems
Human TNFR1, Fc Tag (TN1-H5251) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 100 µG
Catalog # ACRBTN1-H5251100UG
Supplier:  ACRO Biosystems
Frequently Bought Together