Order Entry
Slovakia
ContactUsLinkComponent
Mouse CTLA-4, His Tag (CT4-M52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession NP033973). 1 * 200 µG
Catalog # ACRBCT4-M52H5200UG
Supplier:  ACRO Biosystems
Mouse CTLA-4, His Tag (CT4-M52H5) is expressed from human 293 cells (HEK293). It contains AA Glu 36 - Phe 162 (Accession NP033973). 1 * 200 µG
Catalog # ACRBCT4-M52H5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together