Order Entry
Slovakia
ContactUsLinkComponent
Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 1 mg
Catalog # ACRBLA3-H5255-1MG
Supplier:  ACRO Biosystems
Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accessi on P18627-1). 1 * 1 mg
Catalog # ACRBLA3-H5255-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together