Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus Siglec-15, Fc Tag (SG5-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A2 K5UY47-1). 1 * 100 µG
Catalog # ACRBSG5-C5253100UG
Supplier:  ACRO Biosystems
Cynomolgus Siglec-15, Fc Tag (SG5-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Thr 263 (Accession A0A2 K5UY47-1). 1 * 100 µG
Catalog # ACRBSG5-C5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together