Order Entry
Slovakia
ContactUsLinkComponent
Rabbit 4-1BB, His Tag (41B-R52H9) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Ile 191 (Accession G1SHD7-1). 1 * 100 µG
Catalog # ACRB41B-R52H9100UG
Supplier:  ACRO Biosystems
Rabbit 4-1BB, His Tag (41B-R52H9) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Ile 191 (Accession G1SHD7-1). 1 * 100 µG
Catalog # ACRB41B-R52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together