Order Entry
Slovakia
ContactUsLinkComponent
Canine TIGIT, His Tag (TIT-C52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 208 (Accession F1PKJ0-1). 1 * 100 µG
Catalog # ACRBTIT-C52H7100UG
Supplier:  ACRO Biosystems
Canine TIGIT, His Tag (TIT-C52H7) is expressed from human 293 cells (HEK293). It contains AA Phe 17 - Pro 208 (Accession F1PKJ0-1). 1 * 100 µG
Catalog # ACRBTIT-C52H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together