Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque CD40, Fc Tag (CD0-C5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession NP001 252791.1). 1 * 1 mg
Catalog # ACRBCD0-C5259-1MG
Supplier:  ACRO Biosystems
Rhesus macaque CD40, Fc Tag (CD0-C5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession NP001 252791.1). 1 * 1 mg
Catalog # ACRBCD0-C5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together