Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK29 3). It contains AA Glu 21 - Phe 141 (Accession Q9Y6W8-1 (C136S, C137S)). 1 * 200 µG
Catalog # ACRBICS-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK29 3). It contains AA Glu 21 - Phe 141 (Accession Q9Y6W8-1 (C136S, C137S)). 1 * 200 µG
Catalog # ACRBICS-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together