Order Entry
Slovakia
ContactUsLinkComponent
Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession P31041-1). 1 * 1 mg
Catalog # ACRBCD8-M52H6-1MG
Supplier:  ACRO Biosystems
Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession P31041-1). 1 * 1 mg
Catalog # ACRBCD8-M52H6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together