Order Entry
Slovakia
ContactUsLinkComponent
Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession O54908). 1 * 50 µG
Catalog # ACRBDK1-M52H6-50UG
Supplier:  ACRO Biosystems
Mouse Dkk-1, His Tag (DK1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 272 (Accession O54908). 1 * 50 µG
Catalog # ACRBDK1-M52H6-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together