Order Entry
Slovakia
ContactUsLinkComponent
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab F(ab)sub2sub. 1 * 1 mg
Catalog # ACRBBEB-Y9-1MG
Supplier:  ACRO Biosystems
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab F(ab)sub2sub. 1 * 1 mg
Catalog # ACRBBEB-Y9-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together