Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Human FKBP12 FKBP1A Protein, His Tag,Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contain s AA Met 1 - Glu 108 (Accession AAH05147).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 100 µG
  ACRBFKA-H5122100UG
Recombinat Protein,Human FKBP12 FKBP1A Protein, His Tag,Human FKBP12, His Tag (FKA-H5122) is expressed from E.coli cells. It contain s AA Met 1 - Glu 108 (Accession AAH05147).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 100 µG
  ACRBFKA-H5122100UG
Frequently Bought Together