Order Entry
Slovakia
ContactUsLinkComponent
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH693 19.1). 1 * 100 µG
Catalog # ACRBGIL-H5249100UG
Supplier:  ACRO Biosystems
Human GITR Ligand, His Tag (GIL-H5249) is expressed from human 293 cells (HEK293). It contains AA Gln 50 - Ser 177 (Accession AAH693 19.1). 1 * 100 µG
Catalog # ACRBGIL-H5249100UG
Supplier:  ACRO Biosystems
Frequently Bought Together