Order Entry
Slovakia
ContactUsLinkComponent
Human Glypican 3 (S359F), His Tag, low endotoxin (GP3-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession P51654-1 (S359F)). 1 * 100 µG
Catalog # ACRBGP3-H5223100UG
Supplier:  ACRO Biosystems
Human Glypican 3 (S359F), His Tag, low endotoxin (GP3-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession P51654-1 (S359F)). 1 * 100 µG
Catalog # ACRBGP3-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together