Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human B7-H3 (4Ig), Fc,Avitag (B7B-H82F5) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (A ccession Q5ZPR3-1). 1 * 200 µG
Catalog # ACRBB7B-H82F5200UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H3 (4Ig), Fc,Avitag (B7B-H82F5) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (A ccession Q5ZPR3-1). 1 * 200 µG
Catalog # ACRBB7B-H82F5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together