Order Entry
Slovakia
ContactUsLinkComponent
Recombinat Protein,Human Apolipoprotein A-II APOA2 Protein, His Tag,Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed from E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the C 1 * 100 µG
Catalog # ACRBAPA-H51H6100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Apolipoprotein A-II APOA2 Protein, His Tag,Human Apolipoprotein A-II, His Tag (APA-H51H6) is expressed from E.coli cells. It contains AA Ala 19 - Gln 100 (Accession P02652-1).This protein carries a polyhistidine tag at the C 1 * 100 µG
Catalog # ACRBAPA-H51H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together