Order Entry
Slovakia
ContactUsLinkComponent
Human SIRP alpha, Fc Tag (HPLC-verified) (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession NP001035111). 1 * 1 mg
Catalog # ACRBSIA-H5251-1MG
Supplier:  ACRO Biosystems
Human SIRP alpha, Fc Tag (HPLC-verified) (SIA-H5251) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Ac cession NP001035111). 1 * 1 mg
Catalog # ACRBSIA-H5251-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together