Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque PCSK9, His Tag (PC9-C52H2) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession A8T 666). 1 * 50 µG
Catalog # ACRBPC9-C52H2-50UG
Supplier:  ACRO Biosystems
Rhesus macaque PCSK9, His Tag (PC9-C52H2) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession A8T 666). 1 * 50 µG
Catalog # ACRBPC9-C52H2-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together