Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 25 µG
  ACRBPRN-H82F7-25UG
Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Acc ession Q5THQ0-1). 1 * 25 µG
  ACRBPRN-H82F7-25UG
Frequently Bought Together