Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse CD47, His,Avitag (CD7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accessi on Q61735-1). 1 * 25 µG
Catalog # ACRBCD7-M82E4-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse CD47, His,Avitag (CD7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accessi on Q61735-1). 1 * 25 µG
Catalog # ACRBCD7-M82E4-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together