Order Entry
Slovakia
ContactUsLinkComponent
Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on AAI13679). 1 * 100 µG
Catalog # ACRBPD2-H5220100UG
Supplier:  ACRO Biosystems
Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on AAI13679). 1 * 100 µG
Catalog # ACRBPD2-H5220100UG
Supplier:  ACRO Biosystems
Frequently Bought Together