Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human CD155, His,Avitag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Access ion NP006496.4). 1 * 200 µG
Catalog # ACRBCD5-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD155, His,Avitag (CD5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Access ion NP006496.4). 1 * 200 µG
Catalog # ACRBCD5-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together