Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human B7-H6, His,Avitag (B76-H82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Access ion Q68D85-1). 1 * 200 µG
Catalog # ACRBB76-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H6, His,Avitag (B76-H82E5) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 262 (Access ion Q68D85-1). 1 * 200 µG
Catalog # ACRBB76-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together