Order Entry
Slovakia
ContactUsLinkComponent
Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession P01132-1). 1 * 1 mg
Catalog # ACRBEGF-M5265-1MG
Supplier:  ACRO Biosystems
Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession P01132-1). 1 * 1 mg
Catalog # ACRBEGF-M5265-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together