Biotinylated Human Siglec-3, Avitag,His Tag (CD3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession AAH28152.1). 1 * 25 µG
Catalog # ACRBCD3-H82E7-25UG
Supplier: ACRO Biosystems
Frequently Bought Together